ESD safety shoes, lace-up, Mason Pro Rubber Low
Anbieter: ELTEN
Zertifikate
Über diesen Artikel
ESD safety ankle shoes made from wear resistant breathable cowhide leather with fabric lining, reflective materials, a full padded bellows tongue and a dual density rubber/PU sole ERGO-ACTIVE 2.
- Fabric lining for natural ventilation
- Replaceable insole; orthopaedic insoles possible
- Semi-orthopaedic inlay sole ESD
- Soft fleece ESD insole
- Anatomically-shaped insole and ankle support for improved stability
- TPU scuff cap
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected, for low and high temperatures and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, HI/HRO/SRC, form A
Technische Daten
- Catalog No:
- ELTN7282102/47
- ELTN7282102/46
- ELTN7282102/45
- ELTN7282102/44
- ELTN7282102/43
- ELTN7282102/42
- ELTN7282102/41
- ELTN7282102/40
- ELTN7282103/41
- ELTN7282103/40
- ELTN7282102/48
- ELTN7282101/45
- ELTN7282101/44
- ELTN7282103/48
- ELTN7282101/43
- ELTN7282103/47
- ELTN7282103/46
- ELTN7282101/42
- ELTN7282103/45
- ELTN7282101/41
- ELTN7282103/44
- ELTN7282101/40
- ELTN7282103/43
- ELTN7282103/42
- ELTN7282101/46
- ELTN7282101/48
- ELTN7282101/47
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Technische Daten
Zugehörige Informationen
Frequently Bought Together