ContactUsLinkComponent
VWR®, Laboratory Coat, Fluid-Impervious
VWR®, Laboratory Coat, Fluid-Impervious
Catalog # 113-1117
Supplier:  VWR Collection
undefined
VWR®, Laboratory Coat, Fluid-Impervious
Catalog # 113-1117
Supplier:  VWR Collection
Specifications
  • Clothing Size:
    L
  • Garment application:
    Laboratory
  • Colour:
    White
  • Seq:
    01
  • Pk:
    30
Specifications
About this item
Fluid-impervious lab coats are the cleanest and most durable within the entire VWR® Protection line of apparel.
  • Fluid-impervious material designed to repel water
  • Provides maximum protection
  • Excellent particulate barrier
  • Low-linting
Engineered to optimise barrier protection, cleanliness, and comfort, these garments are appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.
Lab coats feature snap front closure, knit cuffs, and three front pockets.
Certifications: Lab coats are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.