Order Entry
Switzerland
ContactUsLinkComponent
Alcohol 94-96%, AR® ACS (denatured with 5% isopropanol and 5% methanol), Macron Fine Chemicals™
Flammable
Irritant
Health
Alcohol 94-96%, AR® ACS (denatured with 5% isopropanol and 5% methanol), Macron Fine Chemicals™
Catalog # 6183-10
CAS Number:  
Flammable
Irritant
Health
Alcohol 94-96%, AR® ACS (denatured with 5% isopropanol and 5% methanol), Macron Fine Chemicals™
Catalog # 6183-10
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth, PVC-coated
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    94.0 - 96.0%
  • For Laboratory,Research,or Manufacturing Use:
    3.5 - 5.5%
  • Specially Denatured Alcohol Formula 3A (200 proof)(v/v):
    94.0 - 96.0 %
  • Methanol (v/v):
    3.5 - 5.5 %
  • Isopropyl Alcohol (2-propanol)(v/v):
    4.0 - 6.0 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.001 %
  • Specific Gravity at 25°C:
    0.782 - 0.793
  • Water (H₂O):
    ≤0.5 %

Specification Test Results

About this item

 :
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:64-17-5
MDL Number:MFCD00003568