Order Entry
Switzerland
ContactUsLinkComponent
Sodium sulphate, anhydrous ≥99%, Reagent Grade, free-flowing, Fluka™
Sodium sulphate, anhydrous ≥99%, Reagent Grade, free-flowing, Fluka™
Certificates
undefined
Sodium sulphate, anhydrous ≥99%, Reagent Grade, free-flowing, Fluka™

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Solution 10 g to 100 mL (H₂O):
    complying
  • Loss on drying (130°C):
    Max. 0.5 %

Specification Test Results

Related Information

Frequently Bought Together