Order Entry
Switzerland
ContactUsLinkComponent
Potassium bromide concentrate for dilution to 1L, 0.1 M KBr volumetric solution, Fluka™
Potassium bromide concentrate for dilution to 1L, 0.1 M KBr volumetric solution, Fluka™
Katalognr. 32036-1EA
CAS Number:  
Potassium bromide concentrate for dilution to 1L, 0.1 M KBr volumetric solution, Fluka™
Katalognr. 32036-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
Density:1.0 g/cm³ (20 °C)
 :119 g/mol
Storage Temperature:Ambient
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358