

Dichloromethane ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
:75-09-2
:
:CH₂Cl₂
:MFCD00000881
:84.93 g/mol
:39…41 °C (1.013 hPa)
:–97 °C
:1.32…1.33 g/cm³ (20 °C)
:Ambient
:200-838-9
:6.1,III
:Stabilised with amylene
:1593
:GHS08
:GHS07
- Appearance:complying
- Identity (IR):complying
- Assay (GC):Min. 99.80 %
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.02 %
- Free acid (as HCl):Max. 0.001 %
- Free chlorine:complying
- Absorbance at 260 nm:Max. 0.01
- Absorbance at 245 nm:Max. 0.10
- Absorbance at 240 nm:Max. 0.30
- Absorbance at 235 nm:Max. 0.70
- Absorbance:complying
- APHA:Max. 10
- 2-methyl-2-buten (GC):10 - 60 ppm
- Autoignition Temperature:1223 °F
- Contains:amylene as stabilizer
- Refractive Index:n20/D 1.424 (lit.)
Frequently Bought Together