
Acetic acid 1.0 M CH3COOH (1.0N) volumetric solution, Fluka™
Catalog # 34256-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:60.05 g/mol
Boiling Pt:100 °C
Density:1.000 g/cm³
EINECS:200-580-7
:GHS05
Storage Temperature:Ambient
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152