ESD safety sandals, Velcro® fastening, Wellmaxx, Impulse XXT Easy
Anbieter: ELTEN
Zertifikate
Über diesen Artikel
Metal and leather free, super lightweight ESD safety shoes made from microfibre upper, reflective materials, a padded tongue and a dual density PU/PU sole WELLMAXX TRAINERS.
- Fabric lining for natural ventilation
- Full-length inlay sole ESD PRO black
- Soft fleece ESD insole
- Anatomically shaped insole and ankle support for improved stability
- Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
- Protective composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/blue
Certifications: EN 61340-5-1 (ESD), EN ISO 20345; S1, SRC, form A
Technische Daten
- Catalog No:
- ELTN719755/39
- ELTN719755/40
- ELTN719755/41
- ELTN719755/42
- ELTN719755/38
- ELTN719755/36
- ELTN719755/37
- ELTN719755/43
- ELTN719755/44
- ELTN719755/45
- ELTN719755/47
- ELTN719755/48
- ELTN719755/46
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Technische Daten
Frequently Bought Together