NAD+ lithium salt ≥90% (by enzymatic purity), Calbiochem®
Catalog # 481915-1
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity (by Enzymatic Assay):≥90%
Specification Test Results
About this item
Synonyms: β-Nicotinamide-adenine Dinucleotide, Li
:
:669.4 g/mol
Merck Index:14,06344
Storage Temperature:Freezer
CAS Number:64417-72-7
Formula:C₂₁H₂₆N₇O₁₄P₂Li
Storage Temperature:Gefrierschrank