Order Entry
Switzerland
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Silver

QUANTOFIX® Test Strips, Silver

QUANTOFIX® silver, 0 • 1 • 2 • 3 • 5 • 7 • 10 - g/L - Ag⁺, Color reaction of silver, 44×42×126 mm (packaging), 100 Tests, 100
Catalog # MANA91350
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Silver
Catalog # MANA91350
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Silver
QUANTOFIX® Test Strips, Silver

QUANTOFIX® silver, 0 • 1 • 2 • 3 • 5 • 7 • 10 - g/L - Ag⁺, Color reaction of silver, 44×42×126 mm (packaging), 100 Tests, 100

CAS Number:  
Price
SFr. 32.70
Log in for availability
1 * 100 ST
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips are for the rapid and reliable determination of Silver in solutions. The easy dip-and-read-procedure provides a reliable result within 20 seconds.

Specifications

Specifications

  • Chemical method:
    Color reaction of silver
  • Pk:
    100
  • Description:
    QUANTOFIX® silver
  • Dimensions:
    44×42×126 mm (packaging)
  • Tests per kit:
    100 Tests
  • Range:
    0 • 1 • 2 • 3 • 5 • 7 • 10 - g/L - Ag⁺
  • Cat. No.:
    MANA91350
  • Parameters:
    All reagents included
  • Contents:
    100 test strips in a tube, instruction leaflet
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    No
  • Shelf life (months):
    30 (from production)
  • Method:
    Silver - Silver sulfide (yellow → brown)
  • Weight:
    40 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.