Test discs, salinity, Saltesmo
Catalog # MANA90608
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Pk:30
- Description:Saltesmo test discs
- Tests per kit:30
- Range:0 - 5 g/l NaCl
Specifications
About this item
Saltesmo test discs are frequently used for the determination of salt (NaCl) in foodstuffs and in sea water. All reagents are impregnated on the paper, making the test very easy to use even for untrained persons.
- Quick results: just expose to sample, wait and read against multi-stage colour scale
- Always ready to use: no accessories necessary for evaluation
- Simple rapid detection of chloride, bromide and iodide
One simply has to pierce a hole into the disc using the needle supplied with the test and allow the disc to soak in the sample for two minutes. A discoloured zone appears around the hole and the size is proportional to the Chloride concentration in the solution.
Colour change: Red to white