Food standards, Aflatoxin B1 solutions, (CRM), Supelco®
Supplier: Merck
Certificates
About this item
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Jerricans, Narrow Neck, HDPE
Supplier: RIXIUS
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Supplier: EPPENDORF