Order Entry
Switzerland
ContactUsLinkComponent
Heat resistant gloves
Heat resistant gloves
Catalog # JUTEH125A238-W2-9
Supplier:  JUTEC
Heat resistant gloves
Catalog # JUTEH125A238-W2-9
Supplier:  JUTEC

Specifications

  • Glove size:
    9
  • Ambidextrous / hand-specific:
    Ambidextrous
  • Disposable / reusable:
    Reusable
  • Description:
    5-finger gloves
  • Length:
    380 mm
  • EN norm:
    EN 388 (3443), EN 407 (423444)
  • Pk:
    1 PAIR

Specifications

About this item

5-finger gloves made of aluminised preox-aramide fabric.

  • Available as 3-finger or 5-finger gloves or mittens
  • Provide protection against contact heat up to 250 °C (radiant heat up to 1000 °C)
  • Soft, flexible material

For use when working with tools in foundries. Also ideal for glass processing.

Certifications: EN 388 (3443), EN 407 (423444); Cat. III