Order Entry
Switzerland
ContactUsLinkComponent
Furaptra (Mag-Fura-2) tetrapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Furaptra (Mag-Fura-2) tetrapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50035.
Supplier:  Biotium
CAS Number:  
Furaptra (Mag-Fura-2) tetrapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50035.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :587 g/mol
Storage Temperature:Freezer
CAS Number:132319-57-4
Formula:C₁₈H₁₀K₄N₂O₁₁