Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PMAxx™
PMAxx™ dye 20 mM in water
PMAxx™ dye 20 mM in water
PMAxx™ dye 20 mM in water
Catalog # 40069.
Supplier:
Biotium
CAS Number:
Certificates
Expand image
PMAxx™ dye 20 mM in water
Catalog # 40069.
Supplier:
Biotium
CAS Number:
Certificates
Specifications
Pack type:
Vial
Pk:
100 µl
Seq:
1286
Specifications
About this item
Product Details
Documentation
Complementary Products
:
Storage Temperature:
Freezer
Product Details
Documentation
Complementary Products