Order Entry
Switzerland
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 1500 ng/ml counterstain
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 1500 ng/ml counterstain
Catalog # ABNOU0031
Supplier:  Abnova
CAS Number:  
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 1500 ng/ml counterstain
Catalog # ABNOU0031
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070
  • Pk Info:
    2×0.5 ml

Specifications

About this item

 :
 :350.25 g/mol
Storage Temperature:Refrigerator
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681