Supelco® Organic Reference Standard, Thymol solution
Thymol solution, 2000 µg/ml in methanol, Certified reference material, Ampule of 1 ml, 1 mL
Catalog # CRM40188
Supplier: Merck
Jump to:
Product Details & Documents
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311
Specifications
Specifications
- Concentration:2000 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- Description:Thymol solution
- Pk Info:Ampule of 1 ml
- Cat. No.:CRM40188
- M-Clarity quality level:MQ300
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Flash point:9,7 °C - Closed cup
- Molecular weight:150,22
- CAS No.:89-83-8
- Shipping conditions:WICE
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



