Supelco® Cerilliant® Organic Reference Standard, (+\-)-4-Hydroxy-3-methoxymandelic Acid-D₃ (ring-D₃) solution
Catalog # H-091-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- Description:(+\-)-4-Hydroxy-3-methoxymandelic Acid-D₃ (ring-D₃) solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Synonym(s):VMA-D₃ solution
- Molecular weight:201,19
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311