5-Carboxyrhodamine 6G hydrochloride einfaches Isomer ≥95% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
Catalog # 90012.
Supplier: Biotium
CAS Number:
Specifications
- VE:5 mg
- Seq:2210
Specifications
About this item
:
:494.97 g/mol
Storage Temperature:Kühlschrank
Formula:C₂₇H₂₇ClN₂O₅