SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, indapamide, (CRM), Supelco®
Pharmaceutical secondary standard, indapamide, (CRM), Supelco®
Catalog # PHR1660-1G
Supplier:
Merck
CAS Number:
Expand image
Pharmaceutical secondary standard, indapamide, (CRM), Supelco®
Catalog # PHR1660-1G
Supplier:
Merck
CAS Number:
Specifications
Pk:
1 g
CAS No.:
26807-65-8
Description:
Indapamide
Environmentally Preferable:
Description:
Indapamide
Specifications
About this item
Product Details
Product Details