SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, Avobenzone, (CRM), Supelco®
Pk:
1 g
CAS No.:
70356-09-1
Description:
Avobenzone
Environmentally Preferable:
Description:
Avobenzone