Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Potassium Phthalate Monobasic
Supelco® Organic Reference Standard, Potassium Phthalate Monobasic
Catalog # 14659-1G
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Potassium Phthalate Monobasic
Catalog # 14659-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Standards:
    Standard for quantitative NMR, TraceCERT®
  • CAS No.:
    877-24-7
  • Description:
    Potassium phthalate monobasic
  • Storage conditions:
    15...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00013070
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    KHP; Phthalic acid monopotassium salt; Potassium biphthalate; Potassium hydrogen phthalate
  • Melting point:
    295...300 °C (dec.) (lit.)
  • Formula:
    HOOCC₆H₄COOK
  • Molecular weight:
    204,22
  • Flash point:
    197,3 °C - Pensky-Martens closed cup

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.