Supelco® Organic Reference Standard, ʟ-Phenylalanine
40541-100MG
: Merck
:
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:63-91-2
- Description:ʟ-Phenylalanine
- Storage conditions:20...25 °C
- Shipping conditions:STD
- MDL No.:MFCD00064227
- M-Clarity quality level:MQ300
- Synonym(s):(S)-2-Amino-3-phenylpropionic acid
- Melting point:270...275 °C (dec.) (lit.)
- Formula:C₆H₅CH₂CH(NH₂)CO₂H
- Molecular weight:165,19
- Flash point:Not applicable
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.