Supelco® TraceCERT® Organic Reference Standard, ᴅʟ-α-Tocopherol acetate
92359-50MG
: Merck
:
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:7695-91-2
- Description:ᴅʟ-α-Tocopherol acetate
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00072042
- M-Clarity quality level:MQ300
- Synonym(s):Vitamin E acetate; all-rac-α-Tocopheryl acetate
- Molecular weight:472,74
- Density:0,96 g/ml at 20 °C (lit.)
- Flash point:225,5 °C - Closed cup
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.