Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Diuron
Catalog # 16902-50MG
Anbieter:  Merck
CAS-Nummer:  
undefined
Supelco® Organic Reference Standard, Diuron
Catalog # 16902-50MG
Anbieter:  Merck
CAS-Nummer:  

Technische Daten

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    330-54-1
  • Description:
    Diuron
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H302 - H351 - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00018136
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    3-(3,4-Dichlorophenyl)-1,1-dimethylurea
  • Molecular weight:
    233,09
  • Flash point:
    Not applicable

Technische Daten

Über diesen Artikel

  • GHS signal: GHS08, GHS07, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P202 - P260 - P264 - P273 - P301 + P312 - P308 + P313