Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Dimethyl Terephthalate
Supelco® Organic Reference Standard, Dimethyl Terephthalate
Catalog # 07038-1G
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Dimethyl Terephthalate
Catalog # 07038-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Standards:
    Standard for quantitative NMR, TraceCERT®
  • CAS No.:
    120-61-6
  • Description:
    Dimethyl terephthalate
  • Storage conditions:
    15...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008440
  • M-Clarity quality level:
    MQ300
  • Melting point:
    139...141 °C (lit.)
  • Formula:
    C₆H₄-1,4-(CO₂CH₃)₂
  • Molecular weight:
    194,18
  • Flash point:
    151 °C - closed cup

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.