SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, neat, Coumarin, (CRM), TraceCERT®, Supelco®
Organic reference standard, neat, Coumarin, (CRM), TraceCERT®, Supelco®
72609-100MG
:
Merck
:
Organic reference standard, neat, Coumarin, (CRM), TraceCERT®, Supelco®
72609-100MG
:
Merck
:
Pk:
100 mg
CAS No.:
91-64-5
Description:
Coumarin
Description:
Coumarin