Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Drug and metabolite standards, Dehydroepiandrosterone-D5-3-sulfate (DHEAS-D5) (2,2,3,4,4,-D5) sodium salt solution, (CRM)
Supelco® Drug and metabolite standards, Dehydroepiandrosterone-D5-3-sulfate (DHEAS-D5) (2,2,3,4,4,-D5) sodium salt solution, (CRM)
  D-066-1ML
 :  Merck
 :  
undefined
Supelco® Drug and metabolite standards, Dehydroepiandrosterone-D5-3-sulfate (DHEAS-D5) (2,2,3,4,4,-D5) sodium salt solution, (CRM)
  D-066-1ML
 :  Merck
 :  

 

  • Concentration:
    100 µg/ml in methanol (as free sulfate)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    Dehydroepiandrosterone-D5-3-sulfate (DHEAS-D5) (2,2,3,4,4,-D5) sodium salt solution
  • Pk Info:
    ampoule of 1 ml