Supelco® Cerilliant® Organic Reference Standard, Catecholamine Mix 2 (Metanephrines) solution
Catalog # C-110-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml each component in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- Description:Catecholamine mix 2 (Metanephrines) solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- M-Clarity quality level:MQ300
- Synonym(s):Metanephrines solution
- Molecular weight:219,67
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311