Supelco® Cerilliant® Organic Reference Standard, 5α-Dihydrotestosterone-D₃ (16,16,17-D₃) solution
Codice catalogo D-077-1ML
Nome del fornitore: Merck
Numero CAS:
Specifiche
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:79037-34-6
- Description:5α-Dihydrotestosterone-D₃ (16,16,17-D₃) solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Synonym(s):5α-Dihydrotestosterone-d₃ (16,16,17-d₃); 5α-DHT-D3 (16, 16, 17-D₃) solution
- Molecular weight:293,46
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifiche
Informazioni su questo articolo
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311