Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® Cerilliant® Organic Reference Standard, (±)-Normetanephrine-D₃ hydrochloride solution
  N-068-1ML
 :  Merck
 :  
undefined
Supelco® Cerilliant® Organic Reference Standard, (±)-Normetanephrine-D₃ hydrochloride solution
  N-068-1ML
 :  Merck
 :  

 

  • Concentration:
    100 µg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • CAS No.:
    1085333-97-6
  • Description:
    (±)-Normetanephrine-D₃ hydrochloride solution
  • Storage conditions:
    –70 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD17018958
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    222,68
  • Flash point:
    9,7 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

 

 

  • GHS signal: GHS02, GHS06, GHS08
 : P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311