Food standards, Ochratoxin A solution, (CRM), Supelco®
Katalognr. CRM46912
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Concentration:50 µg/ml in benzene: acetic acid (99:1)
- Pk:1
- CAS No.:303-47-9
- Description:Ochratoxin A solution
- Description:Ochratoxin A solution
- Pk Info:ampoule of 1 ml