Order Entry
Switzerland
ContactUsLinkComponent
Food standards, Aflatoxin M1 solution, (CRM), Supelco®
Food standards, Aflatoxin M1 solution, (CRM), Supelco®
Catalog # CRM46319
Supplier:  Merck
CAS Number:  
undefined
Food standards, Aflatoxin M1 solution, (CRM), Supelco®
Catalog # CRM46319
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in acetonitrile
  • Pk:
    1 mL
  • CAS No.:
    6795-23-9
  • Description:
    Aflatoxin M1 solution
  • Description:
    Aflatoxin M1 solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item