ContactUsLinkComponent
HPLC Columns, OPTIMAPACK
 :  HICHROM
HPLC Columns, OPTIMAPACK
 :  HICHROM
  • Catalog No:
  • HISMOPSIL-51002546
  • HISMOP-51002546
  • Carbon load:
  • 20%
  • Endcapped:
  • Yes
  • Yes
  • Surface area:
  • 330 m2/g
  • 330 m2/g
Frequently Bought Together