ContactUsLinkComponent
Electrolyte solution, non-aqueous, Supelco®
Electrolyte solution, non-aqueous, Supelco®
Catalog # 16398-250ML
Supplier:  Merck
CAS Number:  
undefined
Electrolyte solution, non-aqueous, Supelco®
Catalog # 16398-250ML
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    250 mL
  • Description:
    Electrolyte solution, LiCl in ethanol (saturated)
  • Environmentally Preferable:
Specifications
About this item
Lithium chloride solution is used for the determination of trace alkaline earth metals in brines, via chelation ion chromatography, on iminodiacetic acid bonded silica column.
It may also be used along with N,N-dimethylacetamide (DMAC) may be used in size exclusion chromatography of cellulose.