Chemical resistant gloves, uvex rubiflex S NB27S
: UVEX
These chemical protection gloves feature a reinforced cotton interlock lining with a full NBR coating. This makes the gloves suitable for many applications in which chemical, heat or abrasion resistance is required.
- Good resistance to chemicals, acids, alkalis, mineral oils and solvents
- Good heat insulation with reinforced supporting material
- Extremely high flexibility
- Good tactile feel
- Outstanding wearer comfort and ergonomic fit
Typical applications include: Chemical industry, sandblasting, automotive industry, metal working and machine and tool construction.
: EN ISO 374-1:2016 Type A (JKOPT), EN 388:2016 (2121X).
Food contact approved.
Food contact approved.
- Catalog No:
- UVEX9893730
- UVEX9893005
- UVEX9893002
- UVEX9893059
- Model:
- NB27S
- NB27S
- NB27S
- NB27S
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN ISO 374:2016 Type A (JKNOPT), EN 388:2016 (2121X)
- EN ISO 374:2016 Type A (JKNOPT), EN 388:2016 (2121X)
- EN ISO 374:2016 Type A (JKNOPT), EN 388:2016 (2121X)
- EN ISO 374:2016 Type A (JKNOPT), EN 388:2016 (2121X)
- Food contact approved:
- +
- +
- +
- +
Frequently Bought Together