Order Entry
Switzerland
ContactUsLinkComponent
Sleeve protector, Tyvek®
Catalog # SSSA000634
Anbieter:  3S ARBEITSSCHUTZ
undefined
Sleeve protector, Tyvek®
Catalog # SSSA000634
Anbieter:  3S ARBEITSSCHUTZ

Technische Daten

  • Colour:
    White
  • Length:
    600 mm
  • Pk:
    250

Technische Daten

Über diesen Artikel

Sleeve protector made of Tyvek® fabric.

  • Inner seams
  • Elastic at both ends
  • Blue seam at upper end

Length: 60 cm
Colour: White