Safety shoes, lace-up, Uni6, 31629
Supplier: Abeba
Certificates
About this item
Safety shoes are made of breathable textile with pattern.
- Water-repellent
- Lining with silver ions
- Metal-free penetration safety
- Suitable for kitchen
- ORTHOsat sole possible
- Anti-slip TPU outsole
- Sanitized treated
These shoes has soft PU midsole and interchangeable soft comfort insole.
Certifications: EN ISO 20345: 2011 S3, DGUV 112-191, SRC.
Frequently Bought Together