Order Entry
Switzerland
ContactUsLinkComponent
Colour standards, starter set for Lico Spectral colorimeter
Katalognr. 705-1122
Anbieter:  Hach
CAS-Nummer:  
undefined
Colour standards, starter set for Lico Spectral colorimeter
Katalognr. 705-1122
Anbieter:  Hach
CAS-Nummer:  

Technische Daten

  • Pk:
    1 KIT
  • Description:
    Colour standards, starter set for Lico Spectral colorimeter (6×Addista)

Technische Daten

Über diesen Artikel

Calibration solutions for Lico spectral colorimeter.

Delivery: Starter Kit consists of 6×ADDISTA certified colour standard colour solutions, 10× glass round cuvettes, 11 mm, and 10×PMMA rectangular cuvettes, 50 mm