ContactUsLinkComponent
Blue filter for myECL™ imager
Blue filter for myECL™ imager
Catalog # PIER62241
Blue filter for myECL™ imager
Catalog # PIER62241
Specifications
  • Description:
    Blue filter for myECL™ imager
  • Pk:
    1
Specifications
About this item
Thermo Scientific™ 445 to 480 nm bandpass filter is a threaded, 50 mm diameter glass filter. This product is used for the addition of a blue filter to one of two open positions in the myECL Imager for detection of blue-fluorescent dyes and stains.