SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Peroxynitrite Na
Peroxynitrite Na in sodium hydroxide solution
Peroxynitrite Na in sodium hydroxide solution
CAYM81565-1
:
Cayman Chemical
:
Peroxynitrite Na in sodium hydroxide solution
CAYM81565-1
:
Cayman Chemical
:
Pk:
1 mL
Seq:
2070
:
:
85.0 g/mol
:
GHS07
:
Ultra-Low Temperature Freezer (–80°C)
:
14042-01-4
:
ONO₂·Na