Order Entry
Switzerland
ContactUsLinkComponent
 
VISOCOLOR® ECO Analysis Case
  MANA931301
 :  
undefined
VISOCOLOR® ECO Analysis Case
  MANA931301
 :  

 

  • Pk:
    1 SET
  • Description:
    VISOCOLOR® ECO Reagent case
  • Dimensions:
    365×305×168 mm (Including packaging)
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    18
  • Weight:
    1765 g (Including packaging)

 

 

The VISOCOLOR ECO analysis case combines various VISOCOLOR ECO test kits.

  • Evaluation of various parameters for water analysis

Combines test kits for ammonium, carbonate hardness, total hardness, nitrate, nitrite, pH and phosphate in a handy and robust case. It is the ideal mini laboratory for visual water analysis on site.

Hazardous material: Yes

 :

Case, test kits, accessories, instructions.