Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Electrode storage bottles

Electrode storage bottles

Storage bottle for electrodes, 12 ml
Catalog # HACH9260300
Supplier:   Hach
Electrode storage bottles
Catalog # HACH9260300
Supplier:  Hach
Electrode storage bottles

Storage bottle for electrodes, 12 ml

Price
SFr. 9.70
Log in for availability
1 * 1 ST
Product Details & Documents

Electrode storage bottle is used to store pH electrodes.

  • Rubber grommet in lid ensures tight seal with 12 mm electrodes
Specifications
  • Description:
    Storage bottle for electrodes, 12 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.