Order Entry
Switzerland
ContactUsLinkComponent
Ammonium ionophore I - cocktail A, Selectophore™ ion sensor material, Supelco®
Ammonium ionophore I - cocktail A, Selectophore™ ion sensor material, Supelco®
Catalog # 99978-0.1ML-F
Supplier:  Merck
CAS Number:  
Ammonium ionophore I - cocktail A, Selectophore™ ion sensor material, Supelco®
Catalog # 99978-0.1ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, bottomless
  • Pk:
    0,1 mL
  • Seq:
    2010

Specifications

Specification Test Results

  • APPEARANCE (COLOR):
    Brown to Very Dark Brown
  • APPEARANCE (FORM):
    Liquid or Viscous Liquid
  • REFRACTIVE INDEX N20/D:
    1.510 - 1.514
  • INFRARED SPECTRUM:
    CONFORMS TO STRUCTURE
  • RECOMMENDED RETEST PERIOD:
    42 MONTHS

Specification Test Results

About this item

 :