ContactUsLinkComponent
Rotor for Sorvall™ BIOS A centrifuges
Rotor for Sorvall™ BIOS A centrifuges
Catalog # 521-2671
Rotor for Sorvall™ BIOS A centrifuges
Catalog # 521-2671
Specifications
  • Description:
    Fixed-angle rotor Fiberlite™ F6-10x1000 LEX, carbon fiber, 10×1000 ml, 6250 min-1, 12000×g (complete with set of 10 bottles)
  • Pk:
    1