ContactUsLinkComponent
Screw top waste and wash vial for autosampler
Screw top waste and wash vial for autosampler
  PERC09923031
Screw top waste and wash vial for autosampler
  PERC09923031
  • Cap included:
    No
  • Ø×H:
    15×45 mm
  • Capacity:
    4,00 ml
  • Description:
    Clear waste and wash vial
  • Pk:
    1
This screw top waste and wash vial is manufactured from type I borosilicate glass.
 : Meets USP, JP, and EP Pharmacopeia requirements.