ContactUsLinkComponent
Vial locator for GC autosampler
Vial locator for GC autosampler
Catalog # PERCN6101182
Supplier:  PerkinElmer
Vial locator for GC autosampler
Catalog # PERCN6101182
Supplier:  PerkinElmer
Specifications
  • Description:
    Vial locator for GC autosampler
  • Pk:
    1
Specifications
About this item
This is a replacement vial locator for PerkinElmer AutoSystem GC autosamplers.
Included in the GC/MS PSS Injector Starter Kit (N6100447), the GC/MS CAP Injector Starter Kit for Clarus 680/580/480 (N6100448), and the GC/MS 590/690 Capillary Injector Consumable Kit (N6550448).
Compatible with:
Clarus 600
Clarus 400
Clarus 500
Autosystem
Autosystem XL
Clarus 680
Clarus 480
Clarus 590
Clarus 580
Clarus 690