Order Entry
Switzerland
ContactUsLinkComponent
Reagent set, alkalinity (total), VISOCOLOR® ECO
Reagent set, alkalinity (total), VISOCOLOR® ECO
Catalog # MANA931204
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Reagent set, alkalinity (total), VISOCOLOR® ECO
Catalog # MANA931204
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Bromophenol blue;Alkalinity
  • Pk:
    1
  • Description:
    VISOCOLOR® ECO Alkalinity TA, test no. 5-04
  • Dimensions:
    170×115×70 mm (Including packaging)
  • Platform:
    Photometric test kits
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    12
  • Weight:
    127,4 g (Including packaging)
  • For unit:
    PF-12Plus;PF-3 Drinking Water;Advance;PF-3 Pool
  • Tests per kit:
    100
  • Range:
    0.4 - 17.5 °e; 0.10 - 5.00 - mmol/L (H⁺)

Specifications

About this item

Reagents for the determination of alkalinity in water samples.

  • Combines simple implementation with high measurement reliability

For photometric measurement with our compact photometers.

Sea water analysis: Yes

Hazardous material: No

Remarks: Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation.

Delivery: Reagents for 100 tests in a box