Orders LinkContactUsLinkComponent
Total immersion thermometers, solid stem
Total immersion thermometers, solid stem
Supplier:   Amarell
Total immersion thermometers, solid stem
Supplier:  Amarell
Certificates
Total immersion thermometers, solid stem
3 Options Available Below
Product Details & Documents
These glass thermometers are suitable for standard everyday use.
  • White backing
  • Reliable measurements and solid glass
Ø: 5,5 to 7,0 mm.
Product Family Options
Product Information
  • Range
    Division
    Length
    Filling
    Availability
    Price
  • −35...+50 °C
    1,0 °C
    260 mm
    Red liquid
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 27.20
    SFr. 27.20
    1 * 1 ST
       SFr. 27.20
  • −10...+100 °C
    1,0 °C
    260 mm
    Red liquid
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 24.20
    SFr. 24.20
    1 * 1 ST
       SFr. 24.20
  • −10...+150 °C
    1,0 °C
    260 mm
    Red liquid
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 26.70
    SFr. 26.70
    1 * 1 ST
       SFr. 26.70