Order Entry
Switzerland
ContactUsLinkComponent
Potassium permanganate 0.1 mol/l (0.5N) volumetric solution
Potassium permanganate 0.1 mol/l (0.5N) volumetric solution
Catalog # APPC182651.1611
Supplier:  PanReac AppliChem
CAS Number:  
Potassium permanganate 0.1 mol/l (0.5N) volumetric solution
Catalog # APPC182651.1611
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

Specification Test Results

  • Titer at 20 °C:
    0.999 - 1.001
  • Traceability:
    NIST

Specification Test Results

About this item

 :
 :158.03 g/mol
EINECS:231-760-3
UN:3214
ADR:5.1,II
 :GHS03
 :GHS07
 :GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364